Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIKESHI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699441.1 | XP_005699498.1 | 3 - 4 | 651 | 88 | |
GenBank mRNA | KA345874.1 | - | 2 - 3 | 454 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |