Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696027.2 | XP_005696084.2 | 4 - 5 | 328 | 80 |
Gene Symbol: LOC102176170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013973144.2 | XP_013828598.2 | 1 | 555 | 80 |
Gene Symbol: LOC108638494 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018065976.1 | XP_017921465.1 | 1 | 377 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |