Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCLAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043911.1 | XP_017899400.1 | 7 - 8 | 1905 | 64 | |
XM_018043912.1 | XP_017899401.1 | 6 - 7 | 1628 | 64 | ||
XM_018043913.1 | XP_017899402.1 | 6 - 7 | 1752 | 64 | ||
XM_018043914.1 | XP_017899403.1 | 7 - 8 | 1905 | 64 | ||
XM_018043915.1 | XP_017899404.1 | 7 - 8 | 1845 | 64 | ||
XM_018043916.1 | XP_017899405.1 | 7 - 8 | 1905 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |