Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287572.1 | NP_001274501.1 | 3 - 4 | 362 | 86 | |
GenBank mRNA | AB499062.1 | - | 2 - 3 | 198 | 86 | |
AB796345.1 | - | 3 - 4 | 362 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |