Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STMN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689118.2 | XP_005689175.1 | 2 - 3 | 503 | 75 | |
GenBank mRNA | JO635168.2 | - | 3 - 4 | 1718 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |