Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693629.3 | XP_005693686.2 | 6 - 7 | 890 | 93 | |
XM_013972217.2 | XP_013827671.1 | 7 - 8 | 1058 | 93 | ||
XM_013972218.2 | XP_013827672.1 | 6 - 7 | 922 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |