Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013974836.2 | XP_013830290.1 | 2 - 3 | 1963 | 54 | |
XM_018040877.1 | XP_017896366.1 | 3 - 4 | 1823 | 54 | ||
XM_018040878.1 | XP_017896367.1 | 3 - 4 | 1846 | 54 | ||
XM_018040879.1 | XP_017896368.1 | 2 - 3 | 1897 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |