Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690387.3 | XP_005690444.1 | 4 - 5 | 460 | 88 | |
XM_005690388.3 | XP_005690445.1 | 4 - 5 | 394 | 88 | ||
GenBank mRNA | JO412270.2 | - | 4 - 5 | 452 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |