Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285606.1 | NP_001272535.1 | 1 - 2 | 279 | 72 | |
GenBank mRNA | HQ699620.1 | - | 1 - 2 | 280 | 72 | |
JN620334.1 | - | 1 - 2 | 264 | 72 | ||
JN620335.1 | - | 1 - 2 | 264 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |