Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TENT5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056620.1 | XP_017912109.1 | 1 - 2 | 449 | 76 | |
GenBank mRNA | KA315693.1 | - | 1 - 2 | 446 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |