Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PXMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061457.1 | XP_017916946.1 | 4 - 5 | 713 | 78 | |
GenBank mRNA | JO635598.2 | - | 1 - 2 | 66 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |