Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005700757.3 | XP_005700814.3 | 2 - 3 | 816 | 61 | |
GenBank mRNA | JO436725.2 | - | 2 - 3 | 512 | 61 | |
JO644608.2 | - | 2 - 3 | 517 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |