Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUTYH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678542.3 | XP_005678599.2 | 7 - 8 | 873 | 50 | |
XM_005678543.3 | XP_005678600.2 | 7 - 8 | 834 | 50 | ||
XM_005678544.3 | XP_005678601.2 | 7 - 8 | 831 | 50 | ||
XM_013962899.2 | XP_013818353.2 | 7 - 8 | 1252 | 50 | ||
GenBank mRNA | JO603421.2 | - | 10 - 11 | 1221 | 50 | |
KA316438.1 | - | 10 - 11 | 1176 | 50 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |