Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681445.3 | XP_005681502.1 | 3 - 4 | 820 | 55 | |
XM_013964547.2 | XP_013820001.2 | 3 - 4 | 934 | 55 | ||
GenBank mRNA | JO430147.2 | - | 3 - 4 | 827 | 55 | |
JO542782.2 | - | 4 - 5 | 591 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |