Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676337.3 | XP_005676394.2 | 3 - 4 | 541 | 127 | |
XM_018061159.1 | XP_017916648.1 | 3 - 4 | 541 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |