Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678537.3 | XP_005678594.2 | 6 - 7 | 929 | 62 | |
XM_005678538.3 | XP_005678595.2 | 6 - 7 | 897 | 62 | ||
XM_018043596.1 | XP_017899085.1 | 6 - 7 | 929 | 62 | ||
GenBank mRNA | JT648257.1 | - | 6 - 7 | 2651 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |