Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691562.3 | XP_005691619.2 | 2 - 3 | 760 | 64 | |
XM_005691563.3 | XP_005691620.1 | 2 - 3 | 554 | 64 | ||
GenBank mRNA | JT697029.1 | - | 2 - 3 | 538 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |