Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPSNAP3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052540.1 | XP_017908029.1 | 3 - 4 | 80 | ||
GenBank mRNA | JO438003.2 | - | 3 - 4 | 612 | 80 | |
JO610038.2 | - | 3 - 4 | 551 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |