Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2L15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045934.1 | XP_017901423.1 | 3 - 4 | 737 | 76 | |
GenBank mRNA | JO332354.2 | - | 1 - 2 | 210 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |