Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: IL36RN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055334.1 | XP_017910823.1 | 3 - 4 | 185 | 78 | |
GenBank mRNA | KA348203.1 | - | 3 - 4 | 188 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |