Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058546.1 | XP_017914035.1 | 7 - 8 | 1214 | 105 | |
XM_018058547.1 | XP_017914036.1 | 6 - 7 | 1130 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |