Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053658.1 | XP_017909147.1 | 3 - 4 | 379 | 57 | |
XM_018053659.1 | XP_017909148.1 | 3 - 4 | 380 | 57 | ||
GenBank mRNA | KA308244.1 | - | 3 - 4 | 1951 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |