Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSIR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699167.3 | XP_005699224.1 | 5 - 6 | 863 | 66 | |
XM_013975632.2 | XP_013831086.1 | 5 - 6 | 866 | 66 | ||
GenBank mRNA | KA334297.1 | - | 5 - 6 | 875 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |