Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677354.3 | XP_005677411.1 | 2 - 3 | 808 | 73 | |
XM_013976680.2 | XP_013832134.1 | 2 - 3 | 808 | 73 | ||
GenBank mRNA | JO429266.2 | - | 2 - 3 | 811 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |