Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677158.2 | XP_005677215.2 | 2 - 3 | 286 | 71 | |
GenBank mRNA | KA346345.1 | - | 2 - 3 | 284 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |