Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285764.1 | NP_001272693.1 | 4 - 5 | 717 | 106 | |
GenBank mRNA | HM138664.1 | - | 4 - 5 | 878 | 106 | |
HM138665.1 | - | 4 - 5 | 878 | 106 | ||
HM138666.1 | - | 4 - 5 | 878 | 106 | ||
HM852511.1 | - | 4 - 5 | 717 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |