Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFML2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677137.3 | XP_005677194.2 | 2 - 3 | 938 | 71 | |
XM_005677138.3 | XP_005677195.2 | 2 - 3 | 938 | 71 | ||
XM_005677139.3 | XP_005677196.2 | 2 - 3 | 937 | 71 | ||
GenBank mRNA | JO547912.2 | - | 2 - 3 | 912 | 71 | |
JT645285.1 | - | 2 - 3 | 659 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |