Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285611.1 | NP_001272540.1 | 3 - 4 | 308 | 69 | |
XM_005699890.3 | XP_005699947.1 | 3 - 4 | 412 | 69 | ||
GenBank mRNA | EF105410.1 | - | 3 - 4 | 308 | 69 | |
JO427956.1 | - | 3 - 4 | 499 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |