Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005683853.3 | XP_005683910.1 | 6 - 7 | 1969 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |