Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060594.1 | XP_017916083.1 | 3 - 4 | 672 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |