Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018065615.1 | XP_017921104.1 | 7 - 8 | 1280 | 77 | |
GenBank mRNA | JO646258.2 | - | 7 - 8 | 1208 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |