Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691245.3 | XP_005691302.1 | 3 - 4 | 3167 | 93 | |
XM_005691246.3 | XP_005691303.1 | 3 - 4 | 3164 | 93 | ||
GenBank mRNA | JT696609.1 | - | 5 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |