Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674994.3 | XP_005675051.1 | 2 - 3 | 620 | 114 | |
XM_018043725.1 | XP_017899214.1 | 2 - 3 | 647 | 114 | ||
XM_018043773.1 | XP_017899262.1 | 2 - 3 | 1025 | 114 | ||
XM_018043823.1 | XP_017899312.1 | 2 - 3 | 1040 | 114 | ||
GenBank mRNA | JO416306.2 | - | 3 - 4 | 878 | 114 | |
JO455436.2 | - | 1 - 2 | 66 | 114 | ||
JO458498.2 | - | 1 - 2 | 66 | 114 | ||
JO594884.2 | - | 3 - 4 | 2191 | 114 | ||
JT705780.1 | - | 2 - 3 | 545 | 114 | ||
JT737673.1 | - | 2 - 3 | 514 | 114 | ||
KA321199.1 | - | 2 - 3 | 546 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |