Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKS1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677418.2 | XP_005677475.1 | 1 - 2 | 157 | 69 | |
GenBank mRNA | JO446870.2 | - | 1 - 2 | 165 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |