Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050771.1 | XP_017906260.1 | 2 - 3 | 136 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |