Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038729.1 | XP_017894218.1 | 4 - 5 | 1882 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |