Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064758.1 | XP_017920247.1 | 3 - 4 | 897 | 78 | |
GenBank mRNA | JO584058.1 | - | 3 - 4 | 498 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |