Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043437.1 | XP_017898926.1 | 7 - 8 | 984 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |