Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038482.1 | XP_017893971.1 | 1 - 2 | 1906 | 81 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Capra hircus Probe spans exons |