Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681536.3 | XP_005681593.1 | 8 - 9 | 2187 | 133 | |
XM_005681537.3 | XP_005681594.1 | 6 - 7 | 2358 | 133 | ||
XM_005681538.3 | XP_005681595.1 | 7 - 8 | 2054 | 133 | ||
XM_005681539.3 | XP_005681596.1 | 5 - 6 | 1761 | 133 | ||
XM_013964599.2 | XP_013820053.1 | 5 - 6 | 1743 | 133 | ||
XM_013964600.2 | XP_013820054.1 | 5 - 6 | 1775 | 133 | ||
XM_013964601.2 | XP_013820055.1 | 6 - 7 | 2356 | 133 | ||
XM_018049385.1 | XP_017904874.1 | 6 - 7 | 2145 | 133 | ||
XM_018049386.1 | XP_017904875.1 | 7 - 8 | 2457 | 133 | ||
GenBank mRNA | JO429085.2 | - | 1 - 2 | 200 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |