Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLVS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684505.3 | XP_005684562.1 | 3 - 4 | 1198 | 100 | |
XM_018052989.1 | XP_017908478.1 | 3 - 4 | 1378 | 100 | ||
GenBank mRNA | JO636666.2 | - | 3 - 4 | 1743 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |