Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052369.1 | XP_017907858.1 | 4 - 5 | 1031 | 84 | |
GenBank mRNA | JO425315.1 | - | 4 - 5 | 1077 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |