Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C24H18orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039777.1 | XP_017895266.1 | 6 - 7 | 1686 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |