Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NYNRIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054648.1 | XP_017910137.1 | 4 - 5 | 2510 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |