Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MZT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005687650.3 | XP_005687707.1 | 1 - 2 | 188 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |