Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005694759.3 | XP_005694816.1 | 2 - 3 | 316 | 71 | |
XM_013972769.2 | XP_013828223.1 | 2 - 3 | 338 | 71 | ||
XM_018065504.1 | XP_017920993.1 | 2 - 3 | 414 | 71 | ||
XM_018065505.1 | XP_017920994.1 | 2 - 3 | 331 | 71 | ||
GenBank mRNA | JO442205.2 | - | 2 - 3 | 248 | 71 | |
JO447941.2 | - | 2 - 3 | 248 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |