Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005700620.3 | XP_005700677.3 | 4 - 5 | 1357 | 72 | |
GenBank mRNA | JO425258.1 | - | 4 - 5 | 1268 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |