Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAP30L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013965716.2 | XP_013821170.2 | 1 - 2 | 863 | 112 | |
GenBank mRNA | JO385347.2 | - | 3 - 4 | 420 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |