Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013971835.2 | XP_013827289.2 | 3 - 4 | 1057 | 81 | |
XM_018064148.1 | XP_017919637.1 | 3 - 4 | 939 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |