Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676652.3 | XP_005676709.1 | 3 - 4 | 701 | 82 | |
XM_005676653.3 | XP_005676710.1 | 3 - 4 | 627 | 82 | ||
XM_018058549.1 | XP_017914038.1 | 3 - 4 | 802 | 82 | ||
GenBank mRNA | JO426679.1 | - | 3 - 4 | 880 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |